Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Benzocaine, Ph.Eur., RS
Organic reference standard, Benzocaine, Ph.Eur., RS
Catalog # EDQMB0600000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Benzocaine, Ph.Eur., RS
Catalog # EDQMB0600000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    50 mg
  • Standards:
    Reference standard
  • CAS No.:
    94-09-7
  • Description:
    Benzocaine
  • Formula:
    C₉H₁₁NO₂
  • Storage temperature:
    Ambient
  • Description:
    Benzocaine

Specifications

About this item

  • In Monographs 0011 and 1088