Order Entry
Austria
ContactUsLinkComponent
 
Organic reference standard, Beclometasone dipropionate anhydrous, Ph.Eur., RS
  EDQMB0305000
 :  EDQM
 :  
Organic reference standard, Beclometasone dipropionate anhydrous, Ph.Eur., RS
  EDQMB0305000
 :  EDQM
 :  

 

  • Pack type:
    Vial
  • Pk:
    150 mg
  • Standards:
    Reference standard
  • CAS No.:
    5534-09-8
  • Description:
    Beclometasone dipropionate anhydrous
  • Formula:
    C₂₈H₃₇ClO₇
  • Storage temperature:
    Ambient
  • Description:
    Beclometasone dipropionate anhydrous

 

 

  • Method: Monograph only
  • API product type
  • Pharmaceutical applications