Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Amlodipine besilate, Ph.Eur., RS
Organic reference standard, Amlodipine besilate, Ph.Eur., RS
Catalog # EDQMY0000049
Supplier:  EDQM
CAS Number:  
Organic reference standard, Amlodipine besilate, Ph.Eur., RS
Catalog # EDQMY0000049
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    150 mg
  • Standards:
    Reference standard
  • CAS No.:
    111470-99-6
  • Description:
    Amlodipine besilate
  • Formula:
    C₂₆H₃₁ClN₂O₈S
  • Storage temperature:
    Ambient
  • Description:
    Amlodipine besilate

Specifications

About this item

Filled under inert atmosphere protected from humidity.

  • In Monograph 1491
  • Solubility in water: 1 - 10 g/l