Order Entry
Austria
ContactUsLinkComponent
Chromate ion chromatography standard, VHG
Chromate ion chromatography standard, VHG
Catalog # PROOVHG-ICRO-100
CAS Number:  
Chromate ion chromatography standard, VHG
Catalog # PROOVHG-ICRO-100
CAS Number:  

Specifications

  • Concentration:
    1000 mg/l
  • Matrix:
    H₂O
  • Pk:
    100 mL
  • Description:
    Chromate standard

Specifications

About this item

This solution is intended for use as a certified reference material (CRM) and calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.

Certifications: Traceable to NIST and SRM.