Order Entry
Austria
Orders LinkContactUsLinkComponent
pH buffer solutions, colour coded

pH buffer solutions, colour coded

pH buffer solutions, colour coded
Supplier:  REAGECON
Certificates
pH buffer solutions, colour coded

16 Options Available Below

Product Details & Documents

Colour coded pH buffer solutions for the calibration of pH meters.

  • Can be tested at 20 or 25 °C

Certifications: Certified by Reagecon’s ISO 17025 accredited test method and NIST traceable.

Documents

Product Family Options

Product Information

  • pH value
    pH accuracy
    Colour
    Reference temperature
    Pk
    Availability
    Price
  • 4,00
    ±0,01
    Red
    20 °C
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 31,90 €
    31,90 €
    1 * 500 mL
       31,90 €

  • 4,00
    ±0,01
    Red
    25 °C
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 32,90 €
    32,90 €
    1 * 500 mL
       32,90 €

  • 4,00
    ±0,01
    Red
    20 °C
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 46,90 €
    46,90 €
    1 * 1 L
       46,90 €

  • 4,00
    ±0,01
    Red
    25 °C
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 49,30 €
    49,30 €
    1 * 1 L
       49,30 €

  • 4,00
    ±0,01
    Red
    20 °C
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 141,00 €
    141,00 €
    1 * 5 L
       141,00 €

  • 7,00
    ±0,01
    Yellow
    20 °C
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 31,90 €
    31,90 €
    1 * 500 mL
       31,90 €

  • 7,00
    ±0,01
    Yellow
    25 °C
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 32,90 €
    32,90 €
    1 * 500 mL
       32,90 €

  • 7,00
    ±0,01
    Yellow
    20 °C
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 46,90 €
    46,90 €
    1 * 1 L
       46,90 €

  • 7,00
    ±0,01
    Yellow
    25 °C
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 49,30 €
    49,30 €
    1 * 1 L
       49,30 €

  • 7,00
    ±0,01
    Yellow
    20 °C
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 141,00 €
    141,00 €
    1 * 5 L
       141,00 €

  • 10,00
    ±0,01
    Blue
    20 °C
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 31,90 €
    31,90 €
    1 * 500 mL
       31,90 €

  • 10,00
    ±0,01
    Blue
    25 °C
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 32,90 €
    32,90 €
    1 * 500 mL
       32,90 €

  • 10,00
    ±0,01
    Blue
    20 °C
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 46,90 €
    46,90 €
    1 * 1 L
       46,90 €

  • 10,00
    ±0,01
    Blue
    25 °C
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 49,30 €
    49,30 €
    1 * 1 ST
       49,30 €

  • 10,00
    ±0,01
    Blue
    20 °C
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 141,00 €
    141,00 €
    1 * 5 L
       141,00 €

  • 10,00
    ±0,01
    Blue
    25 °C
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 148,00 €
    148,00 €
    1 * 5 L
       148,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.