ContactUsLinkComponent
Cytology swabs, Histobrush®
Cytology swabs, Histobrush®
  PMPC25-2199
undefined
Cytology swabs, Histobrush®
  PMPC25-2199
  • Ø:
    5 mm
  • Description:
    Cytology brushes
  • Length:
    178 mm
  • Pk:
    1.000
These cytology brushes feature tapered tips made out of soft nylon bristles and a long PS handle.
Used for the collection of cervical specimens during pap smear tests, as well as buccal cells, for DNA lineage.
 : ISO 9001:2008, 13485:2003, FDA GMP’s and 510K approved.
 : A certificate of compliance is available upon request.