Order Entry
Austria
ContactUsLinkComponent
 
High performance, manual defrost, −20 °C freezers, TSX
    
 
undefined
High performance, manual defrost, −20 °C freezers, TSX
    

 

  • Catalog No:
  • HERATSX3020EV
  • HERATSX2320EV
  • HERATSX3020FV
  • HERATSX2320FA
  • HERATSX2320FV
  • Model:
  • TSX3020EV
  • TSX2320EV
  • TSX3020FV
  • TSX2320FA
  • TSX2320FV
  • Environmentally Preferable:
  • Energy Efficient, Safer Human and Environmental Health
  • Energy Efficient, Safer Human and Environmental Health
  • Energy Efficient, Safer Human and Environmental Health
  • Energy Efficient, Safer Human and Environmental Health
  • Cat. No.:
  • HERATSX3020EV
  • HERATSX2320EV
  • HERATSX3020FV
  • HERATSX2320FA
  • HERATSX2320FV
  • Display:
  • Microprocessor
  • Microprocessor
  • Microprocessor
  • Microprocessor
  • Microprocessor
  • Defrost:
  • Manual
  • Manual
  • Manual
  • Manual
  • Manual
  • Alarm:
  • Audible and visual
  • Audible and visual
  • Audible and visual
  • Audible and visual
  • Audible and visual
  • Electrical:
  • 208 - 230 V, 50 Hz
  • 230 V, 50 Hz, 16 A
  • 208 - 230 V, 50 Hz
  • 115 V, 60 Hz, 4,3 A
  • 230 V, 50 Hz, 16 A
  • Shelves/drawers:
  • 9 shelves
  • 9 shelves, 45 bins
  • 4 shelves
  • 4 shelves
  • 4 shelves
  • Door type:
  • 1 solid door
  • 1 solid door
  • 1 solid door
  • 1 solid door
  • 1 solid door
  • Plug:
  • CEE 7/7
  • CEE 7/7
  • CEE 7/7
  • NEMA 5-15
  • CEE 7/7
  • Weight:
  • 220 kg
  • 216 kg
  • 188 kg
  • 176 kg
  • 191 kg

 

 

 

Frequently Bought Together