About this item
Thermo Scientific™ TSX Series high-performance pharmacy refrigerators are designed with features that support sample protection and sustainability objectives for the storage of pharmaceuticals, vaccines, chemotherapy and other medical and pharmacy-grade storage requiring 2 to 8 °C. The Series’ V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection. The Series’ V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection. Certified to NSF/ANSI 456 Vaccine Storage standard, the TSX series can help you meet regulations and guidelines from the CDC, World Health Organisation, and other Health Ministries on vaccine storage and handling. The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.
- Triple panel glass door helps maintain optimal internal conditions
- Forced-air circulation for temperature uniformity and fast temperature recovery
- Heat-free defrost for maximum temperature uniformity
- Quiet operation at just 52 dBA
- Keyed on/off switch with easy to use, setpoint security
- Self-closing door with 90° stop to assist with inventory loads
- Built-in remote alarm contacts
V-drive technology: While conventional refrigerant, high-performance refrigerators and freezers use single speed compressors that continually cycle on and off, the TSX Series V-drive runs at variable speeds to adjust cooling performance to the conditions inside and outside of the refrigerator. When combined with the automated tuning control, this variable speed drive optimizes the compressors running speed to the current conditions. When there are frequent door openings, or samples are added to the refrigerator, the control system detects the activity and increases the drive speed to bring temperatures back to the set point quickly. This innovative technology is one of the reasons the TSX Series delivers outstanding door opening recovery (DOR) speed, and more peace of mind for busy laboratories. When conditions are stable, such as when the unit is running overnight or on weekends, the drive runs at a low speed, reducing energy consumption while maintaining a stable temperature for the samples.
Tested to GMP Clean Room Class A/ISO 5 & 6 (ISO EN 14644-1) with easy to clean surfaces support USP and hazardous drug compounding and other clean room environments. Tested and certified to conform with the design and safety standards of UL 471, 61010, and CE Directive 2011/65/EU and 2015/863. Ergonomically designed door handle with a soft-touch grip. The microprocessor controller is angled to help prevent neck strain, Convenient, fully extendable and adjustable basket drawers, Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction. Self-closing door with 90° stop to assist with inventory loads, US EPA ENERGY STAR Certified with up to 40% less energy usage than conventional refrigerant models. ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life.
Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations. Manufactured in an award-winning, zero-waste to landfill facility. Thermo Scientific™ DeviceLink™ Connect ready, enabling you to have a connected experience with your unit so you always and anywhere know the safety of your samples and the performance of your refrigerator or freezer. Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems. Alternate door swing configuration and door types. Extra and alternate wire baskets, shelves, and drawers. Free-standing or factory installed chart recorders. Stainless steel interior finish.
E-Lock adapter kits, 4 to 20 milliamp transmitters and other remote alarm modules. Specialty, regional, and country specific plugs. Industry Leading 120-month compressor warranty (varies by country, please contact us for details). Included Certificate of Calibration to an NIST (National Institute of Standards and Technology, formerly National Bureau of Standards) traceable instrument includes a model number and serial number certificate. Optional Validation Testing Programs include in house and field-based temperature mapping and Installation Qualification (IQ) and Operation Qualification (OQ) programs, Optional Factory Acceptance Test results including QC Test Log Sheets and QC Temperature Graphs.
Specifications
- Catalog No:
- HERATSX1205PV
- HERATSX5005PV
- HERATSX3005PV
- HERATSX2305PV
- Model:
- —
- —
- —
- TSX2305PV
- Environmentally Preferable:
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient, Safer Human and Environmental Health
- Cat. No.:
- HERATSX1205PV
- HERATSX5005PV
- HERATSX3005PV
- HERATSX2305PV
- Temperature:
- 3...7 °C (5 °C)
- 3...7 °C (5 °C)
- 3...7 °C (5 °C)
- 3...7 °C (5 °C)
- Display:
- Microprocessor
- Microprocessor
- Microprocessor
- Microprocessor
- Defrost:
- Automatic
- Automatic
- Automatic
- Automatic
- Alarm:
- Alarm, recorder
- Alarm, recorder
- Alarm, recorder
- Audible and visual
- Shelves/drawers:
- 5 fully extendable and adjustable wire baskets
- 8 fully extendable and adjustable wire baskets
- 6 fully extendable and adjustable wire baskets
- 6 fully extendable and adjustable wire baskets
- Door type:
- 1 Glass door
- Double glass door
- 1 Glass door
- 1 glass swing door
- Refrigerant:
- R290
- R290
- R290
- R290
- Specifications:
- —
- —
- —
- 4 casters; the front 2 are lockable
Bright, LED interior lighting
230 V, 16 A, 50 Hz
- Electrical:
- 208-230 V, 50 Hz
- 208-230 V, 50 Hz
- 208-230 V, 50 Hz
- 208-230 V, 50 Hz
- Plug:
- CEE 7/7
- CEE 7/7
- CEE 7/7
- CEE 7/7
- Weight:
- 151 kg
- —
- 214 kg
- 237 kg
- Shipping weight:
- 176 kg
- —
- 245 kg
- 231 kg