Order Entry
Austria
Orders LinkContactUsLinkComponent
Menaquinone, Reference Standards

Menaquinone, Reference Standards

Supplier:   USP
Menaquinone, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1381119.
  • 1381083.
  • Description:
  • Menaquinone-7
  • Menaquinone-4
  • Synonym(s):
  • Vitamin K2
  • 2-Methyl-3-[(2E,6E,10E)-3,7,11,15-tetramethylhexadeca-2,6,10,14-tetraen-1-yl]naphthalene-1,4-dione
  • CAS No.:
  • 2124-57-4
  • 863-61-6
  • Formula:
  • C₄₆H₆₄O₂

Specifications

Product Family Options

Product Information

  • Standards
    Pk Info
    Pack type
    Pk
    Availability
    Price
  • 1381119.
    Menaquinone-7
    Menaquinone-7
    USP reference standard
    3 x 50 mg
    3
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 414,00 €
    414,00 €
    1 * 3 ST
       414,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1381119.
    Description
    Menaquinone-7
    Synonym(s)
    Vitamin K2
    CAS No.
    2124-57-4
    Formula
    C₄₆H₆₄O₂
  • 1381083.
    Menaquinone-4
    Menaquinone-4
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 367,00 €
    367,00 €
    1 * 50 mg
       367,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1381083.
    Description
    Menaquinone-4
    Synonym(s)
    2-Methyl-3-[(2E,6E,10E)-3,7,11,15-tetramethylhexadeca-2,6,10,14-tetraen-1-yl]naphthalene-1,4-dione
    CAS No.
    863-61-6
    Formula
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.