ContactUsLinkComponent
Multi-parameter meter (pH/ISE/conductivity/DO), bench, inoLab® Multi 9630 IDS
Multi-parameter meter (pH/ISE/conductivity/DO), bench, inoLab® Multi 9630 IDS
Supplier:  WTW
Certificates
undefined
Multi-parameter meter (pH/ISE/conductivity/DO), bench, inoLab® Multi 9630 IDS
Supplier:  WTW
Specifications
  • Catalog No:
  • 665-0487
  • 665-0525
  • Model:
  • inoLab® Multi 9630
  • inoLab® Multi 9630
  • pH range:
  • Depending on sensor
  • Depending on sensor
  • pH calibration:
  • 1- to 5-point
  • 1- to 5-point
  • mV range:
  • Depending on sensor
  • Depending on sensor
  • ISE calibration:
  • 2- to 7-point, adapter required
  • 2- to 7-point, adapter required
  • Conductivity range:
  • Depending on sensor
  • Depending on sensor
  • Conductivity calibration:
  • 1-point
  • 1-point
  • DO concentration:
  • Depending on sensor
  • Depending on sensor
  • DO saturation:
  • Depending on sensor
  • Depending on sensor
  • DO calibration:
  • 1-point
  • 1-point
  • Temperature compensation:
  • All except ORP
  • All except ORP
  • Display:
  • 16:9 colour graphic display
  • 16:9 colour graphic display
  • Power supply:
  • Universal power supply (100 - 240 V, 50/60 Hz)
  • Universal power supply (100 - 240 V, 50/60 Hz)
  • Data logging - log function:
  • Manual or time-controlled
  • Manual or time-controlled
  • Data logging capacity:
  • Manual: 500 data sets; automatic: 10000 data sets
  • Manual: 500 data sets; automatic: 10000 data sets
  • Outputs:
  • USB-A, Mini USB-B
  • USB-A, Mini USB-B
Specifications
Sensors
Product Information
Frequently Bought Together