Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Iopamidol, Reference Standards

 :   USP
Iopamidol, Reference Standards
 :  USP
 
Iopamidol, Reference Standards

 

 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

 

 

 

  • Catalog No:
  • 1344702.
  • 1344724.
  • 1344735.
  • 1344746.
  • Description:
  • Iopamidol
  • Iopamidol Related Compound A
  • Iopamidol Related Compound B
  • Iopamidol Related Compound C
  • Synonym(s):
  • N,N'-Bis-(1,3-dihydroxy-2-propyl)-5-amino-2,4,6-triiodoisophthalamide
  • 5-Glycolamido-N,N'-bis[2-hydroxy-1-(hydroxymethyl)ethyl]-2,4,6-triiodoisophthalamide
  • 4-chloro-N1, N3-bis(1,3-dihydroxypropan-2-yl)-5-(S)-lactamido-2,6-diiodoisophthalamide
  • CAS No.:
  • 60166-93-0
  • 60166-98-5
  • 77868-41-8
  • Formula:
  • C₁₇H₂₂I₃N₃O₈
  • C₁₄H₁₈I₃N₃O₆
  • C₁₆H₂₀I₃N₃O₈

 

Product Family Options

 

  • Description
    Standards
    Pack type
    Pk
     
     
  • Iopamidol
    USP reference standard
    Vial
    200 mg
       
       
    1 * 200 mg
    1 * 200 mg - 406,00 €
    406,00 €
    1 * 200 mg
       406,00 €

     
     
    Specifications
    Cat. No.
    1344702.
    Description
    Iopamidol
    Synonym(s)
    CAS No.
    60166-93-0
    Formula
    C₁₇H₂₂I₃N₃O₈
  • Iopamidol Related Compound A
    USP reference standard
    Vial
    50 mg
       
       
    1 * 50 mg
    1 * 50 mg - 1 180,00 €
    1 180,00 €
    1 * 50 mg
       1 180,00 €

     
     
    Specifications
    Cat. No.
    1344724.
    Description
    Iopamidol Related Compound A
    Synonym(s)
    N,N'-Bis-(1,3-dihydroxy-2-propyl)-5-amino-2,4,6-triiodoisophthalamide
    CAS No.
    60166-98-5
    Formula
    C₁₄H₁₈I₃N₃O₆
  • Iopamidol Related Compound B
    USP reference standard
    Vial
    50 mg
       
       
    1 * 50 mg
    1 * 50 mg - 1 270,00 €
    1 270,00 €
    1 * 50 mg
       1 270,00 €

     
     
    Specifications
    Cat. No.
    1344735.
    Description
    Iopamidol Related Compound B
    Synonym(s)
    5-Glycolamido-N,N'-bis[2-hydroxy-1-(hydroxymethyl)ethyl]-2,4,6-triiodoisophthalamide
    CAS No.
    77868-41-8
    Formula
    C₁₆H₂₀I₃N₃O₈
  • Iopamidol Related Compound C
    USP reference standard
    Vial
    40 mg
       
       
    1 * 40 mg
    1 * 40 mg - 1 230,00 €
    1 230,00 €
    1 * 40 mg
       1 230,00 €

     
     
    Specifications
    Cat. No.
    1344746.
    Description
    Iopamidol Related Compound C
    Synonym(s)
    4-chloro-N1, N3-bis(1,3-dihydroxypropan-2-yl)-5-(S)-lactamido-2,6-diiodoisophthalamide
    CAS No.
    Formula
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.