Beryllium standard solution, for ICP, in HNO₃, CRM, VHG
Supplier: LGC Standards PROMOCHEM
Certificates
About this item
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.
Frequently Bought Together