Orders LinkContactUsLinkComponent
Portable lockout station
Portable lockout station
Portable lockout station, Yellow, 432×171×432 mm
Catalog # BRDY811217
Supplier:   Brady
Portable lockout station
Catalog # BRDY811217
Supplier:  Brady
Portable lockout station
Portable lockout station
Portable lockout station, Yellow, 432×171×432 mm
Price
181,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This portable storage station ensures that lockout devices remain easily accessible and ready for use whenever needed.
  • Made from rugged and rust-proof PP
  • Attachable handle for easy portability
  • Screw slots for permanent wall mounting
  • Lockable, semi-transparent cover protects the contents from exposure to harsh environments
Ordering information: Station is supplied empty (without contents).
Specifications
  • Type:
    Portable lockout station
  • Colour:
    Yellow
  • W×D×H:
    432×171×432 mm