Mag-Bind® stool DNA 96 kit
The Mag-Bind® Stool DNA 96 Kit allows rapid and reliable isolation of high-quality genomic DNA (both pathogen and host DNA) from stool samples.
- Up to 300 μl stool samples can be processed in less than 45 minutes
- System allows for automation after sample lysis
- Two protocols are available depending on the amount of inhibition and downstream sensitivity*
- No organic extraction
The system combines the Mag-Bind® technology with HTR Reagent to eliminate PCR inhibiting compounds such as humic acid from stool samples. Purified DNA is suitable for PCR, restriction digestion, and hybridisation applications. There are no organic extractions thus reducing plastic waste and hands-on time to allow multiple samples to be processed in parallel.
*NOTE: It is recommended to begin with the standard protocol and if issues occur, use the inhibitor rich sample protocol.
- Method / Format:Magnetic beads
- Prep size:Scalable
- Refine target molecule:Total DNA
- Sample type:Stool
- Target molecule:DNA
- Sample size:Up to 300 µl
- Description:Mag-Bind® Stool DNA 96 Kit
Frequently Bought Together