ContactUsLinkComponent
Plug lockout, Stopower™
Plug lockout, Stopower™
Catalog # BRDY065673
Supplier:  Brady
undefined
Plug lockout, Stopower™
Catalog # BRDY065673
Supplier:  Brady
Specifications
  • Type:
    Plug lockout
  • Colour:
    Red
  • W×D×H:
    27,5×18,7×44,7 mm
  • Pk:
    1
Specifications
About this item
This plug hugger is ideal for office equipment and other frequently locked out cords.
  • For use on 2 or 3 pronged polarised plugs
  • Cord cannot be plugged in with Stopower in place
  • Device cannot be removed without a key