Order Entry
Austria
Orders LinkContactUsLinkComponent
Levothyroxine, Reference Standards

Levothyroxine, Reference Standards

Supplier:   USP
Levothyroxine, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1365000.
  • 1365065.
  • 1365043.
  • Description:
  • Levothyroxine
  • Levothyroxine Sodium
  • Levothyroxine Peak Identication Mixture
  • CAS No.:
  • 51-48-9
  • 25416-65-3 (hydrate)
  • Formula:
  • C₁₅H₁₁I₄NO₄
  • C₁₅H₁₂I₄NNaO₅
  • UN No.:
  • 1230

Specifications

Product Family Options

Product Information

  • Standards
    Pk Info
    Pack type
    Pk
    Availability
    Price
  • 1365000.
    Levothyroxine
    Levothyroxine
    USP reference standard
    Vial
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 418,00 €
    418,00 €
    1 * 500 mg
       418,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1365000.
    Description
    Levothyroxine
    CAS No.
    51-48-9
    Formula
    C₁₅H₁₁I₄NO₄
    UN No.
  • 1365065.
    Levothyroxine Sodium
    Levothyroxine Sodium
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 418,00 €
    418,00 €
    1 * 50 mg
       418,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1365065.
    Description
    Levothyroxine Sodium
    CAS No.
    25416-65-3 (hydrate)
    Formula
    C₁₅H₁₂I₄NNaO₅
    UN No.
  • 1365043.
    Levothyroxine Peak Identication Mixture
    Levothyroxine Peak Identication Mixture
    USP reference standard
    3 x 5 mL 1 mg/mL
    Ampule
    3
    Log in for availability
    Log in for availability
    1 * 3 ST
    1 * 3 ST - 1 410,00 €
    1 410,00 €
    1 * 3 ST
       1 410,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1365043.
    Description
    Levothyroxine Peak Identication Mixture
    CAS No.
    Formula
    UN No.
    1230
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.