Order Entry
Austria
Orders LinkContactUsLinkComponent
Isosorbide, Reference Standards

Isosorbide, Reference Standards

Supplier:   USP
Isosorbide, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1353102.
  • 1353113.
  • 1352008.
  • Description:
  • Diluted Isosorbide Mononitrate
  • Diluted Isosorbide Mononitrate Related Compound A
  • Isosorbide 75% solution
  • Synonym(s):
  • 1,4:3,6-Dianhydro-D-glucitol 2-nitrate in lactose
  • CAS No.:
  • 16051-77-7
  • 16106-20-0
  • 652-67-5

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1353102.
    Diluted Isosorbide Mononitrate
    Diluted Isosorbide Mononitrate
    USP reference standard
    Vial
    750 mg
    Log in for availability
    Log in for availability
    1 * 750 mg
    1 * 750 mg - 466,00 €
    466,00 €
    1 * 750 mg
       466,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1353102.
    Description
    Diluted Isosorbide Mononitrate
    Synonym(s)
    CAS No.
    16051-77-7
  • 1353113.
    Diluted Isosorbide Mononitrate Related Compound A
    Diluted Isosorbide Mononitrate Related Compound A
    USP reference standard
    Vial
    150 mg
    Log in for availability
    Log in for availability
    1 * 150 mg
    1 * 150 mg - 1 400,00 €
    1 400,00 €
    1 * 150 mg
       1 400,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1353113.
    Description
    Diluted Isosorbide Mononitrate Related Compound A
    Synonym(s)
    1,4:3,6-Dianhydro-D-glucitol 2-nitrate in lactose
    CAS No.
    16106-20-0
  • 1352008.
    Isosorbide 75% solution
    Isosorbide 75% solution
    USP reference standard
    Ampule
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 369,00 €
    369,00 €
    1 * 1 g
       369,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1352008.
    Description
    Isosorbide 75% solution
    Synonym(s)
    CAS No.
    652-67-5
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.