Order Entry
Austria
Orders LinkContactUsLinkComponent
Indinavir, Reference Standards

Indinavir, Reference Standards

Supplier:   USP
Indinavir, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1339178.
  • 1339189.
  • Description:
  • Indinavir
  • Indinavir System Suitability
  • CAS No.:
  • 180683-37-8
  • 180683-37-8
  • Formula:
  • C₃₆H₄₉N₅O₅
  • C₃₆H₄₉N₅O₅

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1339178.
    Indinavir
    Indinavir
    USP reference standard
    Vial
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 333,00 €
    333,00 €
    1 * 100 mg
       333,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1339178.
    Description
    Indinavir
    CAS No.
    180683-37-8
    Formula
    C₃₆H₄₉N₅O₅
  • 1339189.
    Indinavir System Suitability
    Indinavir System Suitability
    USP reference standard
    Vial
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 643,00 €
    643,00 €
    1 * 100 mg
       643,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1339189.
    Description
    Indinavir System Suitability
    CAS No.
    180683-37-8
    Formula
    C₃₆H₄₉N₅O₅