Glyburide, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1295505.
- 1295516.
- 1295527.
- Description:
- Glyburide
- Glyburide Related Compound A
- Glyburide Related Compound B
- Synonym(s):
- 4-[2-(5-chloro-2-methoxybenzamido)ethyl]benzenesulfonamide
- methyl-[[4-[2-[(5-chloro-2-methoxybenzoyl)-amino]ethyl]phenyl]sulfonyl]carbamate
- CAS No.:
- 10238-21-8
- 16673-34-0
- 21165-77-5
- Formula:
- C₂₃H₂₈ClN₃O₅S
- C₁₆H₁₇ClN₂O₄S
- C₁₈H₁₉ClN₂O₆S
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1295505.GlyburideSpecifications:
More
Specifications
Cat. No.1295505.DescriptionGlyburideSynonym(s)CAS No.10238-21-8FormulaC₂₃H₂₈ClN₃O₅S - 1295516.Glyburide Related Compound AGlyburide Related Compound AUSP reference standardVial25 mgLog in for availability1 * 25 mg1 240,00 €Specifications:
More
Specifications
Cat. No.1295516.DescriptionGlyburide Related Compound ASynonym(s)4-[2-(5-chloro-2-methoxybenzamido)ethyl]benzenesulfonamideCAS No.16673-34-0FormulaC₁₆H₁₇ClN₂O₄S - 1295527.Glyburide Related Compound BGlyburide Related Compound BUSP reference standardVial25 mgLog in for availability1 * 25 mg1 260,00 €Specifications:
More
Specifications
Cat. No.1295527.DescriptionGlyburide Related Compound BSynonym(s)methyl-[[4-[2-[(5-chloro-2-methoxybenzoyl)-amino]ethyl]phenyl]sulfonyl]carbamateCAS No.21165-77-5FormulaC₁₈H₁₉ClN₂O₆S



