Orders LinkContactUsLinkComponent
Universal valve lockout, base + arm
 :   Brady
Universal valve lockout, base + arm
 :  Brady
Universal valve lockout, base + arm
Universal valve lockout, base + arm
These universal lockout devices are a modular system which enables users to lock out valves of different types and sises.
  • Flexibility and security
  • Available in two different sizes
Max. shackle diameter: 9 mm
Min. shackle clearance: 20 mm
Product Family Options
  • Type
    Colour
  • Small universal valve lockout with base and arm, max. handle thickness: 15,24 mm, max. valve handle width: 25,4 mm
    Red
    1 * 1 ST
    1 * 1 ST - 135,00 €
    135,00 €
    1 * 1 ST
       135,00 €
  • Large universal valve lockout with base and arm, max. handle thickness: 27,94 mm, max. valve handle width: 40,64 mm
    Red
    1 * 1 ST
    1 * 1 ST - 157,00 €
    157,00 €
    1 * 1 ST
       157,00 €