Order Entry
Austria
ContactUsLinkComponent
Human recombinant Inositol Monophosphatase 1 (from E. coli)
Human recombinant Inositol Monophosphatase 1 (from E. coli)
Catalog # PRSI91-214
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Inositol Monophosphatase 1 (from E. coli)
Catalog # PRSI91-214
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Environmentally Preferable:
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Inositol Monophosphatase 1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    32.3 kD
  • Sequence:
    Met1-Asp277
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl, pH 7.25. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological assays

Specifications

About this item

Inositol Monophosphatase 1 (IMPA1) belongs to the inositol monophosphatase family. IMPA1 is responsible for the provision of inositol required for synthesis of phosphatidylinositol and polyphosphoinositides, IMPA1 can use myo-inositol-1,3-diphosphate, myo-inositol-1,4-diphosphate, scyllo-inositol-phosphate, glucose-1-phosphate, glucose-6-phosphate, fructose-1-phosphate, beta-glycerophosphate, and 2-AMP as substrates. IMPA1 has been implicated as the pharmacological target for lithium action in brain. IMPA1 shows magnesium-dependent phosphatase activity and is inhibited by therapeutic concentrations of lithium. In addition, IMPA1 plays a improtant role in intracellular signal transduction.

  • Fusion-Tag: N-6 His tag

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.