Human recombinant IL10 (from E. coli)
PRSI90-498
:
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:His Tag
- Storage conditions:Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
- Protein synonyms:Interleukin-10|Cytokine Synthesis Inhibitory Factor|CSIF
- Protein/peptide name:IL10
- Purity:>95% (SDS-PAGE)
- Sequence:19-178
- Formulation:Lyophilized from 0.2um-filtered solution in PBS (pH 7.4).
Interleukin-10 (IL-10) is a cytokine produced by activated Th2 cells, B cells, keratinocytes and monocytes/macrophages. In vitro murine and human IL-10 inhibits cytokine synthesis by Th1 cells, natural killer cells and monocytes/macrophages. Several studies have suggested the potential application of IL-10 as an anti-inflammatory agent in the treatment of septic shock and as an immunosuppressive agent in certain T cell mediated autoimmune diseases.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.