Order Entry
Austria
ContactUsLinkComponent
Human recombinant BMP2 (from CHO cells)
Human recombinant BMP2 (from CHO cells)
Catalog # PRSI90-372
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant BMP2 (from CHO cells)
Catalog # PRSI90-372
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    Stable for at least 1 year after receipt when stored at −20 °C. Working aliquots are stable for up to 3 months when stored at −20 °C.
  • Biological activity:
    Shows the biological function of the BMP-2 moiety and exerts a prolonged circulating half-life caused by the modified Fc domain.
  • Protein synonyms:
    Bone Morphogenetic Protein 2|BMP2
  • Protein/peptide name:
    BMP2
  • Purity:
    >98% (SDS-PAGE)
  • Sequence:
    289-396
  • Formulation:
    Lyophilized from 0.2um-filtered solution in PBS.

Specifications

About this item

Bone morphogenetic protein 2 (BMP-2) is a member of the BMP subgroup of the TGF-beta superfamily. It plays a dominant role in embryonic dorsalventral patterning, organogenesis, limb bud formation and bone formation and regeneration. BMP-2 signals through heterodimeric complexes composed of a type I receptor (Activin RI, BMP-RIA or BMP-RIB) and a type II receptor (BMP-RII or Activin RIIB). BMP-2 induces chondrocyte proliferation, endochondral bone formation, longitudinal bone growth and bone and cartilage repair. It induces ectopic bone formation or calcification by promoting osteogenic and chondrogenic differentiation in mesenchymal cells, stem cells and vascular smooth muscle cells. It also promotes the maintenance and repair of colonic epithelium, suppresses neuronal dopamine synthesis and release, induces apoptosis in medulloblastoma cells and is required for cardiac contractility.

Fusion-Tag: Fc Tag

This recombinant proteins is for research use only.