Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Serum
- Species:Human
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:HSA|serum albumin|HAS|serum of albumin
- Protein/peptide name:Albumin
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:66.5 kD
- Sequence:Asp25-Leu609
- Formulation:Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Human serum albumin (HSA), the most abundant protein in human blood plasma, is essential for maintaining osmotic pressure. It is produced in the liver, consists of a single polypeptide chain, with 585 amino acids with 17 tyrosil residues and one tryptophan located in position 214. HSA is organised in three domains, I, II and III, each consisting of two subdomains, A and B. In the physiological states, HSA occurs in two forms – the non-modified HSA with a free thiol group of Cys-34, and the modified (oxidised) form (oHSA), also called human mercaptoalbumin (HMA) and human nonmercaptoalbumin (HNA), respectively. HMA and HNA are in equilibrium, depending on the redox state of Cys-34, and their ratio also varies depending on age and the diseased state. HSA functions primarily as a carrier protein for drugs, steroids, fatty acids, and thyroid hormones, and plays a role in stabilising extracellular fluid volume.
This recombinant protein can be used for biological assays. For research use only.