Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:N-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Liver tissue 2D-page spot 71B|Antioxidant enzyme B166|Peroxisomal antioxidant enzyme|TPx type VI|PRDX5|AOEB166|Alu corepressor 1|Thioredoxin reductase|Prx-V|Peroxiredoxin V|Thioredoxin peroxidase PMP20|Peroxiredoxin-5|PLP
- Protein/peptide name:Peroxiredoxin 5
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:17.9 kD
- Sequence:Met53-Leu214
- Formulation:Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Specifications
About this item
Peroxisomes are essential organelles that participate in multiple important metabolic processes, including the beta -oxidation of fatty acids, plasmalogen synthesis, and the metabolism of reactive oxygen species (ROS). Peroxiredoxins is overexpressed in breast cancer tissues to a great extent suggesting that they has a proliferative effect and may be related to cancer development or progression. Peroxiredoxin 5 (PRDX5) is a thioredoxin peroxidase that belongs to the atypical 2-Cys class of the TSA/ahpC family of peroxiredoxins. PRDX5 is a widely expressed mitochondrial antioxidant enzyme that reduces hydrogen peroxide, alkyl hydroperoxides, and peroxynitrite. In human cells, this enzyme is present in the cytosol, mitochondria, peroxisomes, and nucleus.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.