Order Entry
Austria
ContactUsLinkComponent
Human recombinant IL-15 R alpha (from Cells)
Human recombinant IL-15 R alpha (from Cells)
Catalog # PRSI92-530
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IL-15 R alpha (from Cells)
Catalog # PRSI92-530
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Environmentally Preferable:
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    CD215|MGC104179|alpha|IL-15RA|interleukin-15 receptor subunit alpha|IL-15R-alpha|IL-15 receptor subunit alpha|CD215 antigen|IL15RA|interleukin 15 receptor
  • Protein/peptide name:
    IL-15 R alpha
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    45.5 kD
  • Sequence:
    Ile31-Thr205
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

Interleukin 15 Receptor alpha (IL-15R alpha) is a transmembrane glycoprotein that plays a pleiotropic role in immune development and function, including the positive maintenance of lymphocyte homeostasis. IL-15R alpha chain can bind soluble IL-15 and “transpresent” cytokine to the cells, allowing them to respond to IL-15. Soluble IL-15R alpha can function as a specific high-affinity IL-15 antagonist. The soluble IL-15/IL-15R alpha complexes exhibit a strong agonistic activity which is mediated through membrane-bound IL-15 receptor beta and gamma heterodimers and enables signalling to cells.

Fusion-Tag: C-Fc tag

This recombinant protein can be used for biological assays. For research use only.