Order Entry
Austria
ContactUsLinkComponent
Human Recombinant EIF4E (from E. coli)
Human Recombinant EIF4E (from E. coli)
Catalog # PRSI92-253
Supplier:  ProSci Inc.
CAS Number:  
Human Recombinant EIF4E (from E. coli)
Catalog # PRSI92-253
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Eukaryotic translation initiation factor 4E|eIF-4F 25 kDa subunit|EIF4EL1|eIF-4E|EIF4E|EIF4F|mRNA cap-binding protein
  • Protein/peptide name:
    EIF4E
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    25.1 kD
  • Sequence:
    Met1-Val217
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Eukaryotic translation initiation factor 4E is a 217 amino acids protein that belongs to the eukaryotic initiation factor 4E family. eIF4F is a multi-subunit complex, the composition of which varies with external and internal environmental conditions. It is composed of at least EIF4A, EIF4E and EIF4G1/EIF4G3. EIF4E is also known to interact with other partners.It can recognize and bind the 7-methylguanosine-containing mRNA cap during an early step in the initiation of protein synthesis and facilitates ribosome binding by inducing the unwinding of the mRNAs secondary structures.

This recombinant protein can be used for biological assays. For research use only.