Order Entry
Austria
ContactUsLinkComponent
Human recombinant UBE2V1 (from E. coli)
Human recombinant UBE2V1 (from E. coli)
Catalog # PRSI92-165
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant UBE2V1 (from E. coli)
Catalog # PRSI92-165
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    UEV1|UBE2V|P/OKcl.19|CROC-1|TRAF6-Regulated IKK Activator 1 Beta Uev1A|Ubiquitin-Conjugating Enzyme E2 Variant 1|UEV-1|Uev1a|CROC1|UBE2V1
  • Protein/peptide name:
    UBE2V1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    17.5 kD
  • Sequence:
    Ala2-Asn147
  • Formulation:
    Supplied as a 0.2 um filtered solution of 50mM HEPES, 100mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Ubiquitin-Conjugating Enzyme Variant 1a (UBE2V1) is a member of the Ubiquitin-conjugating (E2) enzyme family. The E2 catalytic core domain of UBE2V1 lacks an active site cysteine residue, rendering it catalytically inactive on its own. However, in the cytoplasm UBE2V1 is able to form a catalytically active complex with UBE2N/Ubc13, which mediates the synthesis Lys63-linked Ubiquitin chains and is required for NF-kappa B activation. UBE2V1 is required for UBE2N (Ubc13)/UBE2V1 Complex-dependent Lys63-linked Ubiquitin chain formation. More specifically, UBE2V1 orients the Ubiquitin molecule to favor linkage at Lys63 via a non-covalent interaction with the Ubiquitin molecule. The UBE2V1-UBE2N heterodimer catalyses the synthesis of non-canonical poly-ubiquitin chains that are linked through Lys63. This type of poly-ubiquitination activates IKK and does not seem to involve protein degradation by the proteasome. UBE2V1 plays a role in the activation of NF-kappa-B mediated by IL1B, TNF, TRAF6, and TRAF2. It mediates transcriptional activation of target genes. UBE2V1 also controls the progress through the cell cycle and differentiation, the error-free DNA repair pathway and contributes to the survival of cells after DNA damage.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.