Order Entry
Austria
ContactUsLinkComponent
 
Human recombinant Peroxiredoxin 3 (from E. coli)
  PRSI92-157
 :  
Human recombinant Peroxiredoxin 3 (from E. coli)
  PRSI92-157
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    Protein MER5 homolog|AOP1|Peroxiredoxin III|AOP-1|PRDX3|HBC189|Thioredoxin-Dependent Peroxide Reductase Mitochondrial|Antioxidant Protein 1|Prx-III|Peroxiredoxin-3
  • Protein/peptide name:
    Peroxiredoxin 3
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    21.6 kD
  • Sequence:
    Pro63-Gln256
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl,pH8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

Thioredoxin-Dependent Peroxide Reductase Mitochondrial (PRDX3) is an enzyme that belongs to the AhpC/TSA family. Human and mouse PRDX3 genes are highly conserved, and they map to the regions syntenic between mouse and human chromosomes. Human PRDX3 protein has an antioxidant function and is localised in the mitochondrion. PRDX3 is involved in redox regulation of the cell. PRDX3 protects radical-sensitive enzymes from oxidative damage by a radical-generating system. It acts synergistically with MAP3K13 to regulate the activation of NF-kappa-B in the cytosol.

  • This recombinant protein can be used for biological assays. For research use only.

This recombinant protein can be used for biological assays. For research use only.