Human recombinant UBE2V2 (from E. coli)
Catalog # PRSI92-156
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:UEV2|MMS2 Homolog|EDPF-1|DDVIT1|Enterocyte Differentiation-Associated Factor 1|DDVit 1|Ubiquitin-Conjugating Enzyme E2 Variant 2|UBE2V2|Enterocyte Differentiation-Promoting Factor 1|EDPF1|MMS2|EDAF-1|Vitamin D3-Inducible Protein
- Protein/peptide name:UBE2V2
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:18.5 kD
- Sequence:Met1-Asn145
- Formulation:Supplied as a 0.2 um filtered solution of 50mm HEPES,150mM NaCl, pH 7.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Ubiquitin-Conjugating Enzyme E2 Variant 2 (UBE2V2) is an enzyme that belongs to the ubiquitin-conjugating enzyme family. UBE2V2 can be detected in the placenta, colon, liver, and skin. It forms a heterodimer with UBE2N. The UBE2V2/UBE2N heterodimer catalyses the synthesis of non-canonical poly-ubiquitin chains and which leads to protein degradation by the proteasome. UBE2V2 mediates transcriptional activation of target genes. It plays a role in the control of progress through the cell cycle and differentiation. It also plays a role in the error-free DNA repair pathway and contributes to the survival of cells after DNA damage.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.