Order Entry
Austria
ContactUsLinkComponent
 
Human recombinant PPP1CC (from E. coli)
  PRSI92-040
 :  
Human recombinant PPP1CC (from E. coli)
  PRSI92-040
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N, C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    PP-1G|Serine/Threonine-Protein Phosphatase PP1-Gamma Catalytic Subunit|Protein Phosphatase 1C Catalytic Subunit|PPP1CC
  • Protein/peptide name:
    PPP1CC
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    40.2 kD
  • Sequence:
    Met1-Lys323
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 1mM DTT, pH 8.0 . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

Serine/Threonine-Protein Phosphatase PP1-Υ Catalytic Subunit (PPP1CC) is a member of the PPP phosphatase family. It is essential for cell division, participates in the regulation of glycogen metabolism, muscle contractility and protein synthesis. PPP1CC colocalizes with SPZ1 in the nucleus, with URI1 at mitochondrion, Rapidly exchanges between the nucleolar, nucleoplasmic and cytoplasmic compartments. As a cofactor, PPP1CC binds one iron ion and one manganese ion per subunit.. In addition, PPP1CC may play an important role in dephosphorylating substrates such as the postsynaptic density-associated Ca2+/calmodulin dependent protein kinase II.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N, C-6 His tag