Order Entry
Austria
ContactUsLinkComponent
Mouse recombinant VEGFA (from P. pichia)
Mouse recombinant VEGFA (from P. pichia)
  PRSI91-944
 :  
Mouse recombinant VEGFA (from P. pichia)
  PRSI91-944
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    P. pichia
  • Species:
    Mouse
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    VEGF164|VEGF 164|VEGFA164|VEGFA|VEGF-A|Vascular permeability factor|VPF|Vascular endothelial growth factor A
  • Protein/peptide name:
    VEGFA
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    19.27 kD
  • Sequence:
    Ala27-Arg190
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    used for biological assays

 

 

Mouse Vascular endothelial growth factor (VEGF or VEGF­A), is a potent mediator of both angiogenesis and vasculogenesis in the fetus and adult. It is a member of the PDGF/VEGF growth factor family that is characterized by a cystine knot structure formed by eight conserved cysteine residues. Alternately spliced isoforms of 120, 164 and 188 aa found in mouse. VEGF binds the type I transmembrane receptor tyrosine kinases VEGF R1 (also called Flt­1) and VEGF R2 (Flk­/KDR) on endothelial cells.Although affinity is highest for binding to VEGF R1, VEGF R2 appears to be the primary mediator of VEGF angiogenic activity. VEGF is required during embryogenesis to regulate the proliferation, migration, and survival of endothelial cells.It may play a role in increasing vascular permeability during lactation, when increased transport of molecules from the blood is required for efficient milk protein synthesis.

  • This recombinant protein can be used for biological assays. For research use only.