Order Entry
Austria
ContactUsLinkComponent
Human recombinant ERBB2 (from cells)
Human recombinant ERBB2 (from cells)
Catalog # PRSI91-939
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant ERBB2 (from cells)
Catalog # PRSI91-939
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-Fc tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    MLN19|Tyrosine kinase-type cell surface receptor HER2|TKR1|ERBB2|CD340|Metastatic lymph node gene 19 protein|Her2|Receptor tyrosine-protein kinase erbB-2|Proto-oncogene Neu|NGL
  • Protein/peptide name:
    ERBB2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    96.5 kD
  • Sequence:
    Thr23-Thr652
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    used for biological assays

Specifications

About this item

Human epidermal growth factor receptor 2 (HER2) is a type of membrane glycoprotein, and belongs to the epidermal growth factor (EGF) receptor family. HER2 plays a key role in development, cell proliferation and differentiation. HER2 has been reported to associate with malignancy and a poor prognosis in numerous carcinomas, including breast, prostate, ovarian, lung cancers and so on. HER2 is activated by dimerization and not activated by EGF, TGF-alpha and amphiregulin. Interaction with PTK6 increases its intrinsic kinase activity.It is heterodimer with EGFR, ERBB3 and ERBB4. HER2 associates with the 5'-TCAAATTC-3' sequence in the PTGS2/COX-2 promoter and activates its transcription. It implicated in transcriptional activation of CDKN1A and the function of the protein involves STAT3 and SRC. And also it involved in the transcription of rRNA genes by RNA Pol I and enhances protein synthesis and cell growth.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-Fc tag