Human Recombinant CD27 (from Cells)
Catalog # PRSI91-931
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-Fc-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:CD27 antigen is also known as CD27L receptor|T-cell activation antigen CD27|T14 and TNFRSF7.|Tumor necrosis factor receptor superfamily member 7|TNFRSF7
- Protein/peptide name:CD27
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:47.2 kD
- Sequence:Thr21-Ile192
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM Tris,150mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
CD27 antigen is also known as CD27L receptor, T-cell activation antigen CD27, Tumor necrosis factor receptor superfamily member 7, T14 and TNFRSF7. In humans, it is encoded by the CD27 gene. CD27 is a single-pass type I membrane protein with 3 TNFR-Cys repeats. It is a member of the TNF-receptor superfamily and is required for generation and long-term maintenance of T cell immunity. It binds to ligand CD70, and plays a key role in regulating B-cell activation and immunoglobulin synthesis. It plays a role in survival of activated T-cells and apoptosis through association with SIVA1.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc-6 His tag