Order Entry
Austria
ContactUsLinkComponent
Human recombinant IL10 (from Cells)
Human recombinant IL10 (from Cells)
Catalog # PRSI91-880
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant IL10 (from Cells)
Catalog # PRSI91-880
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    IL10A|RP11-262N9.1|IL-10|Cytokine synthesis inhibitory factor|TGIF|MGC126450|MGC126451|Interleukin-10|IL10|CSIF
  • Protein/peptide name:
    IL10
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    19.6 kD
  • Sequence:
    Ser19-Asn178
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    used for biological assays

Specifications

About this item

Interleukin 10(IL10), also known as cytokine synthesis inhibitory factor (CSIF),is a secreted protein and belongs to the IL-10 family. IL-10 is secreted by many activated hematopoietic cell types as well as hepatic stellate cells, keratinocytes, and placental cytotrophoblasts . IL-10 is an anti-inflammatory TH2 cytokine that has a critical role in limiting the immune response to pathogens to prevent host damage. As IL-10 in produced in several T helper populations, it is proposed that it provides a feedback loop to limit the effector functions of macrophages and DCs on T cells. Once expressed, IL-10 signals through the IL-10 receptor (IL-10R) to activate STAT3. As IL-10 is a strong inhibitor of inflammation, it has become a viable biomarker for various diseases and conditions as well as a therapeutic molecule for certain conditions. In addition to elevated levels in parasitic infection, high expression levels of IL-10 are also found in retroviral infections inducing immunodeficiency. The immunosuppressive properties of IL-10 suggest a possible clinical use of IL-10 in suppressing rejections of grafts after organ transplantations

  • This recombinant protein can be used for biological assays. For research use only.