- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Collectin Kidney Protein 1|COLEC11|Collectin-11|CL-K1
- Protein/peptide name:Collectin 11
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:27.14 kD
- Sequence:Gln26-Met271
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Collectin-11 is a secreted protein that belongs to the COLEC10/COLEC11 family. Collectin-11 contains one C-type lectin domain and one collagen-like domain. Collectins play important roles in the innate immune system by binding to carbohydrate antigens on microorganisms, facilitating their recognition and removal. Collectin-11 binds to various sugars including fucose and mannose, but does not bind to glucose, N-acetylglucosamine and N-acetylgalactosamine. It has a higher affinity for fucose compared to mannose. Collectin-11 binds lipopolysaccharides (LPS). It also involved in fundamental development serving as a guidance cue for neural crest cell migration. Defects in Collectin-11 are the cause of 3MC syndrome type 2 (3MC2).
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag