Human recombinant B3GNT1 (from Cells)
Catalog # PRSI91-704
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Poly-N-Acetyllactosamine Extension Enzyme|I-Beta-1|UDP-GlcNAc:BetaGal Beta-1|B3GNT1|B3GNT6|3-N-Acetylglucosaminyltransferase 1|3-N-Acetylglucosaminyltransferase|N-Acetyllactosaminide Beta-1|iGnT
- Protein/peptide name:B3GNT1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:43.4 kD
- Sequence:Asp43-Cys415
- Formulation:Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
N-Acetyllactosaminide beta -1,3-N-Acetylglucosaminyltransferase (B3GNT1) is a member of the beta -1,3-N-Acetylglucosaminyltransferase family. B3GNT1 is a single-pass type II membrane protein and widely expressed in many tissues. B3GNT1 can initiate the synthesis or the elongation of the linear poly-N-acetyllactosaminoglycans. B3GNT1 is essential for the synthesis of poly-N-acetyllactosamine, a determinant for the blood group i antigen. It can initiate the synthesis or the elongation of the linear poly-N-acetyllactosaminoglycans.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag