Order Entry
Austria
ContactUsLinkComponent
Human recombinant G6PD (from cells)
Human recombinant G6PD (from cells)
  PRSI91-652
 :  
Human recombinant G6PD (from cells)
  PRSI91-652
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    G6PD|Glucose-6-Phosphate 1-Dehydrogenase
  • Protein/peptide name:
    G6PD
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    60.2 kD
  • Sequence:
    Ala2-Leu515
  • Formulation:
    Supplied as a 0.2 um filtered solution of PBS, pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

Glucose-6-Phosphate 1-Dehydrogenase (G6PD) is a cytosolic enzyme that belongs to the glucose-6-phosphate dehydrogenase family. G6PD participates in the pentose phosphate pathway that supplies reducing energy to cells by maintaining the level of the co-enzyme nicotinamide adenine dinucleotide phosphate (NADPH). G6PD produces pentose sugars for nucleic acid synthesis and main producer of NADPH reducing power. NADPH in turn maintains the level of glutathione in these cells that helps protect the red blood cells against oxidative damage. It is notable in humans that G6PD is remarkable for its genetic diversity. G6PD deficiency may cause neonatal jaundice, acute hemolysis, or severe chronic non-spherocytic hemolytic anemia.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag