Order Entry
Austria
ContactUsLinkComponent
Human recombinant PRPS2 (from cells)
Human recombinant PRPS2 (from cells)
Catalog # PRSI91-645
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant PRPS2 (from cells)
Catalog # PRSI91-645
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Ribose-Phosphate Pyrophosphokinase 2|PRPS2|Phosphoribosyl Pyrophosphate Synthase II|PPRibP|PRS-II
  • Protein/peptide name:
    PRPS2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    35.8 kD
  • Sequence:
    Pro2-Leu318
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Ribose-Phosphate Pyrophosphokinase 2 (PRPS2) is a phosphoribosyl pyrophosphate synthetase that belongs to the ribose-phosphate pyrophosphokinase family. PRPS2 is a homodimer. The active form is probably an hexamer composed of three homodimers. PRPS2 catalyzes the synthesis of phosphoribosylpyrophosphate (PRPP) that is essential for nucleotide synthesis. PRPS2 catalyzes the synthesis of 5-phosphoribosyl 1-pyrophosphate from ATP and D-ribose 5-phosphate. In addition, PRPS2 plays a central role in the synthesis of purines and pyrimidines.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag