Human Recombinant Growth hormone (from E. coli)
Catalog # PRSI91-586
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:GH-N|Somatotropin|Growth hormone 1|GH|Growth hormone|GH1|Pituitary growth hormone
- Protein/peptide name:Growth Hormone
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:22.1 kD
- Sequence:Phe27-Phe217
- Formulation:Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Growth hormone (GH), also known as somatotropin, is a member of a family of growth factors. It plays an important role in growth control. Its major role in stimulating body growth is to stimulate the liver and other tissues to secrete IGF-1. GH includes prolactin, placental lactogens, proliferins, and somatolactin. It is synthesized primarily by somatotropes in the anterior pituitary and is stored in secretory granules. It stimulates both the differentiation and proliferation of myoblasts. It also stimulates amino acid uptake and protein synthesis in muscle and other tissues.
- This recombinant protein can be used for biological assays. For research use only.