Order Entry
Austria
ContactUsLinkComponent
Mouse recombinant IL10 (from E. coli)
Mouse recombinant IL10 (from E. coli)
Catalog # PRSI91-582
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant IL10 (from E. coli)
Catalog # PRSI91-582
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    IL-10|Cytokine synthesis inhibitory factor|Il10|Interleukin-10|CSIF
  • Protein/peptide name:
    IL10
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    18.9 kD
  • Sequence:
    Ser19-Ser178
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Mouse Il10 is the prototypic member of the IL-10 cytokine family, including IL-10, IL-19, IL-20, IL-22 (IL-TIF), IL-24 and IL-26. Many viruses encode viral members of the IL-10 family, such as Epstein-Barr virus (EBV) and human cytomegalovirus (HCMV).Its main function is inhibiting the synthesis of a number of cytokines, including IFN-gamma, IL-2, IL-3, TNF and GM-CSF produced by activated macrophages and by helper T-cells. Although human and mouse IL-10 are 81% identical at the nucleotide and amino acid level, mouse IL-10 is species-specific and does not act on human cells. Interestingly, Human IL-10 is active on mouse cells.

  • This recombinant protein can be used for biological assays. For research use only.