Order Entry
Austria
ContactUsLinkComponent
Human recombinant Secreted Phosphoprotein 1 (from cells)
Human recombinant Secreted Phosphoprotein 1 (from cells)
Catalog # PRSI91-494
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Secreted Phosphoprotein 1 (from cells)
Catalog # PRSI91-494
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Nephropontin|SPP-1|Bone Sialoprotein 1|Urinary Stone Protein|OPN|BNSP|Secreted Phosphoprotein 1|Uropontin|SPP1|Osteopontin
  • Protein/peptide name:
    Secreted Phosphoprotein 1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    34.75 kD
  • Sequence:
    Ile17-Asn314
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological Assays

Specifications

About this item

Secreted Phosphoprotein 1 (SPP1) is a secreted multifunctional glycoprotein. Its putative functions include roles in bone metabolism, immune regulation, wound healing, cell survival, and tumor progression. Based on gene structure and chromosomal location, SPP1 is a member of the small integrin-binding ligand N-linked glycoprotein (SIBLING) family that also includes bone sialoprotein (BSP), dentin matrix protein 1 (DMP1), dentin sialophosphoprotein (DSPP), enamelin (ENAM), and matrix extracellular phosphoglycoprotein (MEPE). SPP1 is expressed in bone, although it is also expressed in other tissues. SPP1 acts as a cytokine that is involved in enhancing production of interferon-gamma and interleukin-12 and reducing production of interleukin-10. It is essential in the pathway that leads to type I immunity. Osteopontin has been implicated as an important factor in bone remodeling. Specifically, research suggests it plays a role in anchoring osteoclasts to the mineral matrix of bones. The fact that SPP1 interacts with multiple cell surface receptors which are ubiquitously expressed makes it an active player in many physiological and pathological processes including wound healing, bone turnover, tumorigenesis, inflammation and ischemia. Therefore, manipulation of plasma Osteopontin levels may be useful in the treatment of autoimmune diseases, cancer metastasis, osteoporosis and some forms of stress.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag