Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Plasma Protease C1 Inhibitor|C1 Inh|C1IN|C1-Inhibiting Factor|C1 Inhibitor|C1Inh|SERPING1|Serpin G1|C1NH|C1 Esterase Inhibitor
- Protein/peptide name:Serpin G1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:53.9 kD
- Sequence:Asn23-Ala500
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
The Human Serpin superfamily consists of at least 35 members that target not only serine proteases, but also selected cysteine proteases and non-protease proteins. As protease inhibitors, serpins have an array of functions including regulating blood clotting, the complement pathway, extracellular matrix remodeling, and cell motility. Serpin G1 is a serine protease inhibitor protein. It is the largest member among the serpin class of proteins. Remarkably, Serpin G1 has a 2-domain structure, unlike most family members. The C-terminal serpin domain is similar to other serpins, and this part of Serpin G1 provides the inhibitory activity. The N-terminal domain is not essential for Serpin G1 to inhibit proteinases and has no similarity to other proteins. The main function of Serpin G1 is the inhibition of the complement system to prevent spontaneous activation. Serpin G1 is an acute phase protein and circulates in blood at levels of around 0.25g/L, whose levels rise 2-fold during inflammation. Although named after its complement inhibitory activity, Serpin G1 also inhibits proteinases of the fibrinolytic, clotting, and kinin pathways. Most notably, Serpin G1 play a potentially crucial role in regulating important physiological pathways including complement activation, blood coagulation, fibrinolysis and the generation of kinins. It is also the most important physiological inhibitor of fXIIa, chymotrypsin and plasma kallikrein.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag