Human recombinant CLIC2 (from E. coli)
Catalog # PRSI91-246
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Chloride Intracellular Channel Protein 2|CLIC2|XAP121
- Protein/peptide name:CLIC2
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:30.5 kD
- Sequence:Met1-Ser247
- Formulation:Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 100mM NaCl, 1mM DTT, 20% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Chloride Intracellular Channel Protein 2 (CLIC2) is a critical component of all living cells; it regulatescellular traffic of Chloride ion and it can be inserted into membranes anf form chloride ion channels. Membrane insertion seems to be redox-regulated and may occur only under oxydizing conditions, channel activity depends on the pH. CLIC2 is involved in regulating membrane potential and organic solute transport. CLIC2 modulates the activity of RYR2 and inhibits Calcium influx. CLIC2 can be detected in the adult brain, liver, lung, heart, stomach, spleen and testis. It is expressed in fetal liver and adult skeletal muscle. CLIC2 is a potential candidate for one of many diseases linked to Xq28.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag