Human recombinant SCO1 (from E. coli)
Catalog # PRSI91-239
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-GST tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:SCOD1|Protein SCO1 Homolog Mitochondrial|SCO1
- Protein/peptide name:SCO1
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:20.14 kD
- Sequence:Gly132-Ser300
- Formulation:Lyophilized from a 0.2 um filtered solution of 50mM PB, 1mM DTT, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Protein SCO1 Homolog, Mitochondrial (SCO1) is a member of the SCO1/2 family. SCO1 has a homodimer structure. SCO1 is located in mitochondrion and is highly expressed in muscle, heart, and brain. It is characterized by high rates of Oxidative Phosphorylation (OxPhos). SCO1 is thought to play a important role in cellular copper homeostasis, mitochondrial redox signaling and insertion of copper into the active site of COX. The defects of SCO1 can result in Mitochondrial Complex IV Deficiency (MT-C4D). A disorder of the mitochondrial respiratory chain has heterogeneous clinical manifestations, ranging from isolated myopathy to severe multisystem disease affecting several tissues and organs.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-GST tag