Order Entry
Austria
ContactUsLinkComponent
Human recombinant HRSP12 (from E. coli)
Human recombinant HRSP12 (from E. coli)
Catalog # PRSI91-236
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant HRSP12 (from E. coli)
Catalog # PRSI91-236
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    PSP|HRSP12|14.5 kDa Translational Inhibitor Protein|Heat-Responsive Protein 12|UK114 Antigen Homolog|Ribonuclease UK114|p14.5
  • Protein/peptide name:
    HRSP12
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    16.7 kD
  • Sequence:
    Met1-Leu137
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 100mM NaCl, 1mM DTT, 10% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

Heat-Responsive Protein 12 (HRSP12) is an endoribonuclease that belongs to the Rut family. HRSP12 is found mainly in the human adult kidney and liver and is responsible for inhibiting protein translation by cleaving mRNA. HRSP12 only cleaves phosphodiester bonds in single-stranded RNA and inhibits cell-free protein synthesis. The levels of both mRNA and protein are markedly reduced in heptatocellular tumors and in human hepatoma cell lines compared with normal liver tissues. Moreover the levels of HRSP12 are different depending on the grade of the tumor. This had led to the suggestion that HRSP12 may be an important biomarker for heptatic carcinoma.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-6 His tag