Order Entry
Austria
ContactUsLinkComponent
 
Human recombinant IFN gamma (from E. coli)
  PRSI91-012
 :  
Human recombinant IFN gamma (from E. coli)
  PRSI91-012
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Biological activity:
    Specific Activity is greater than 1.5 x 107 IU/mg as determined in a viral resistance assay using VSV-WISH cells.
  • Gene ID:
    3458
  • Protein synonyms:
    Interferon Gamma|IFN-Gamma|IFNG|Immune Interferon
  • Protein/peptide name:
    IFN gamma
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    16.88 kD
  • Sequence:
    Gln24-Gln166
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

 

 

IFN gamma is the major interferon produced by mitogenically or antigenically stimulated lymphocytes. It is structurally different from type I interferon and its major activity is immunoregulation. It has been implicated in the expression of class II histocompatibility antigens in cells that do not normally produce them, leading to autoimmune disease. Interferon gamma is produced mainly byT-cells and natural killer cells activated by antigens, mitogens, or alloantigens. It is produced by lymphocytes expressing the surface antigens CD4 and CD8. IFN gamma synthesis is induced by IL-2, FGF-basic, and EGF.

  • This recombinant protein can be used for biological assays. For research use only.