Order Entry
Austria
ContactUsLinkComponent
Human recombinant FABP2 (from E. coli)
Human recombinant FABP2 (from E. coli)
Catalog # PRSI96-290
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant FABP2 (from E. coli)
Catalog # PRSI96-290
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    200 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Biological activity:
    The binding affinity of Recombinant Human FABP2/I‑FABP for the synthetic ligand cis-parinaric acid has been measured by fluorescence titration. Half-maximal fluorescence of 3 μM Recombinant Human FABP2/I‑FABP is achieved with approximately 3 μM cis‑paranaric acid.
  • Protein synonyms:
    I-FABP|FABP2|FABPI
  • Protein/peptide name:
    FABP2
  • Purity:
    >98% as determined by SDS-PAGE.
  • Molecular weight:
    16 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot

Specifications

About this item

Fatty acid-binding protein 2 (FABP2), is also known as Fatty acid-binding protein, intestinal (FABPI), Intestinal-type fatty acid-binding protein (I-FABP). FABP2 belongs to the calycin superfamily and Fatty-acid binding protein (FABP) family. FABP2 / FABPI is expressed in the small intestine and at much lower levels in the large intestine and is highest expressed in the jejunum. FABP are thought to play a role in the intracellular transport of long-chain fatty acids and their acyl-CoA esters. FABP2 is probably involved in triglyceride-rich lipoprotein synthesis. FABP2 binds saturated long-chain fatty acids with a high affinity, but binds with a lower affinity to unsaturated long-chain fatty acids. FABP2 may also help maintain energy homeostasis by functioning as a lipid sensor.