Human recombinant Tdo2 (from E. coli)
Catalog # PRSIXW-RP3263
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Environmentally Preferable:
- Protein synonyms:TRPO|TO|TDO|TPH2|Tryptophan 2|Tryptamin 2|3-dioxygenase
- Protein/peptide name:Tdo2
- Purity:>95%
- Molecular weight:46.7 kDa (Calculated)
- Tested applications:ELISA, Western Blot, MS
Specifications
About this item
The tetramer binds two molecules of protoheme IV. It is a homotetramer involved in tryptophan catabolism. There is a broad specificity towards tryptamine and derivatives including D- and L-tryptophan, 5-hydroxytryptophan and serotonin.
- This recombinant protein can be used for WB, ELISA, MS.
10 mM Tris, pH 8.0, 0.1% Triton X-100, 0.002% NaN
Store in working aliquots at -80°C. Avoid freeze/thaw cycles.