SARS M protein SARS M protein peptide (14 aa near N-terminus)
PRSI3527P
:
- Pk:50 µG
- Species:SARS M protein
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:14 aa near N-terminus
- Protein/peptide name:SARS M protein Peptide (14 aa near N-terminus)
- Tested applications:Blocking
SARS matrix peptide is used for blocking the activity of the SARS matrix antibody.